missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Axygen™ 384 Deep Well Assay Plates

Product Code. 11320115 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11320115 Clear Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11320115 Supplier Corning™ Supplier No. P384240SQCSI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen™ polypropylene assay plates are used for sample collection, in vitro growth chambers and long term storage. With excellent chemical resistance they can withstand temperatures down to -80°C.

  • Excellent chemical resistance
  • Centrifugation to 4,000 rcf
  • Diamond shaped wells allow complete sample recovery
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • ANSI/SLAS 1-2004: Microplates - Footprint dimensions
  • Withstand temperatures to -80°C
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid No
Array 8 x 12
Color Clear
Product Type 384 Deep-well Assay Plate
Material Polypropylene
No. of Wells 384
Sterility Sterile
Total Volume (Metric) Well 240 μL
Well Shape Square, Conical Bottom (V-well)
For Use With (Application) Sample collection, in vitro growth chambers, long term storage
Max. RCF 4000 x g
Temperature Range (Metric) Down to -80 °C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.