missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Polypropylene Microtubes with White Screw Cap

Product Code. 12559027 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
0.5 mL
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12559027 0.5 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12559027 Supplier Axygen Supplier No. SCT050SSWS
  • ONLINE EXCLUSIVE PRICE
    £234.00
    £228.02 / Pack of 500
    Save £5.98
    2% Off
Estimated Shipment: 08-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Cap Style Screw Cap
Closure Color White
Capacity (Metric) 0.5 mL
Material Polypropylene
Includes Screw Cap, O-ring
No. per Pack 4000Pack
Packaging 4000Pack
Sterility Sterile
Graduated No
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.