missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Universal Fit Micropipette Tips

Product Code. 12511308 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Tip Style:
Beveled
Genomic/Wide Orifice
Low Retention, Beveled
Low Retention, Universal
Universal
Quantity:
10,000/Cs.
4800/Cs.
Unit Size:
Pack of 10000
Pack of 4800
Pack of 960
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Color Tip Style Quantity unitSize
12511308 Non-sterile Yellow Beveled 4800/Cs. Pack of 4800
12655876 Non-sterile Clear Beveled 4800/Cs. Pack of 4800
12713119 Non-sterile Clear Genomic/Wide Orifice 4800/Cs. Pack of 4800
11314255 Non-sterile Clear Genomic/Wide Orifice 4800/Cs. Pack of 960
12501298 Non-sterile Clear Low Retention, Beveled 4800/Cs. Pack of 4800
12501308 Sterile Clear Beveled 4800/Cs. Pack of 4800
13197384 Non-sterile Yellow Low Retention, Universal 4800/Cs. Pack of 4800
11314275 Non-sterile Yellow Low Retention, Universal 4800/Cs. Pack of 960
11324275 Sterile Yellow Low Retention, Universal 4800/Cs. Pack of 960
12786619 Sterile Yellow Low Retention, Universal 4800/Cs. Pack of 4800
11334275 Non-sterile Yellow Universal 4800/Cs. Pack of 960
12796619 Non-sterile Yellow Universal 4800/Cs. Pack of 4800
12726729 Sterile Yellow Universal 4800/Cs. Pack of 4800
11344275 Sterile Yellow Universal 4800/Cs. Pack of 960
12551348 Non-sterile Yellow Universal 10,000/Cs. Pack of 10000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 12511308 Supplier Axygen™ Supplier No. TR222YSTK
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Axygen™ Universal Fit Pipette Tips are compatible with most research-grade single- and multi-channel micropipettes, and are certified to be DNase-/RNase-free and nonpyrogenic.

  • RNase-/DNase free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Available in a variety of sizes
  • Available with a variety of tip styles including beveled, fine point, and wide bore
  • Available sterile/non-sterile
  • Available in bulk, racked, or stack rack packaging
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Clean Claims Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free
For Use With (Application) Pipetting Viscous materials, genomic DNA, or delicate cell cultures.
Format Stack Rack
Tip Style Beveled
Packaging 96 x 5 Racks/Stack, 10 Stacks/Cs.
Quantity 4800/Cs.
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 10, 50, 100μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.