missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PCR Tubes with 0.5 mL Flat Cap

Product Code. 11371974 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
Orange
Red
Violet
Yellow
Product Type:
Maxymum Recovery PCR Tube
Thin Wall PCR Tube
Quantity:
1000/Pk.
1000/Pk., 10Pk./Cs.
Unit Size:
Pack of 1000
Pack of 10000
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type Quantity unitSize
11371974 Red Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
12786869 Red Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
12736869 Blue Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
11341974 Blue Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
12726869 Assorted Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
11301984 Assorted Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
12756869 Green Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
11351974 Green Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
12766869 Clear Maxymum Recovery PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
11390145 Clear Maxymum Recovery PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
12746869 Clear Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
11331974 Clear Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
11361974 Orange Thin Wall PCR Tube 1000/Pk. Pack of 1000
12776869 Orange Thin Wall PCR Tube 1000/Pk. Pack of 10000
12796869 Violet Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
11381974 Violet Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
12716879 Yellow Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 10000
11391974 Yellow Thin Wall PCR Tube 1000/Pk., 10Pk./Cs. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 11371974 Supplier Axygen™ Supplier No. PCR05R

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thin and consistent wall thickness for precise thermal transfer

Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.

  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured from polypropylene

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Color Red
Volume (Metric) 0.5 mL
Quantity 1000/Pk., 10Pk./Cs.
Closure Material Polypropylene
For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Shape Conical
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Format Tube
Product Type Thin Wall PCR Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.