missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PCR Tubes with 0.2 mL Dome Cap
GREENER_CHOICE

Product Code. 11370145 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
Orange
Red
Spectrum
Violet
White
Yellow
Product Type:
Maxymum Recovery, Domed Cap PCR Tubes
Thin Wall, Domed Cap PCR Tube
Unit Size:
Pack of 1000
Pack of 10000
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type unitSize
11370145 Clear Maxymum Recovery, Domed Cap PCR Tubes Pack of 1000
11311924 Assorted Thin Wall, Domed Cap PCR Tube Pack of 1000
12756849 Assorted Thin Wall, Domed Cap PCR Tube Pack of 10000
11351914 Blue Thin Wall, Domed Cap PCR Tube Pack of 1000
12766849 Blue Thin Wall, Domed Cap PCR Tube Pack of 10000
10820721 White Thin Wall, Domed Cap PCR Tube Pack of 10000
11341914 White Thin Wall, Domed Cap PCR Tube Pack of 1000
12796849 Clear Maxymum Recovery, Domed Cap PCR Tubes Pack of 10000
12786849 Green Thin Wall, Domed Cap PCR Tube Pack of 10000
11361914 Green Thin Wall, Domed Cap PCR Tube Pack of 1000
11371914 Orange Thin Wall, Domed Cap PCR Tube Pack of 1000
12706859 Orange Thin Wall, Domed Cap PCR Tube Pack of 10000
12716859 Red Thin Wall, Domed Cap PCR Tube Pack of 10000
11381914 Red Thin Wall, Domed Cap PCR Tube Pack of 1000
12565617 Spectrum Thin Wall, Domed Cap PCR Tube Pack of 1000
11380145 Spectrum Thin Wall, Domed Cap PCR Tube Pack of 10000
12726859 Violet Thin Wall, Domed Cap PCR Tube Pack of 10000
11391914 Violet Thin Wall, Domed Cap PCR Tube Pack of 1000
12736859 Yellow Thin Wall, Domed Cap PCR Tube Pack of 10000
11301924 Yellow Thin Wall, Domed Cap PCR Tube Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 11370145 Supplier Axygen™ Supplier No. PCR02DLC

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thin and consistent wall thickness for precise thermal transfer

Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.

  • PCR tubes with 0.2mL volume
  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Color Clear
Volume (Metric) 0.2 mL
Quantity 1000/Pk., 10Pk./Cs.
Closure Material Polypropylene
For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Shape Conical
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Format Tube
Product Type Maxymum Recovery, Domed Cap PCR Tubes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.