Learn More
Axygen™ Microtube Racks: 80-Well Axyracks
Ideal for storage and handling of PCR/Micro/Screw cap tubes
Brand: Axygen R-80-AF
This item is not returnable.
View return policy
Description
- Holds eighty 1.5/2.0mL micro and screw cap tubes
- Polypropylene; resistant to most lab chemicals and solvents
- For cryogenic storage of tubes
- Conical bottom wells for superior stability of conical tubes
- Each well numbered for ease of sample identification
- Frosted side writing panel
- Available in a variety of fluorescent and non-fluorescent colors
- Packaging: 25 Racks/Cs.