missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Microplate Sealing Film and Tapes

Product Code. 11336254 Shop All Axygen Scientific Products
Change view
Click to view available options
Material:
Aluminum
Litto or Ethylene or Vinyl
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
CyclerSeal Sealing Film Roll
Heat Seal Film
Sealing Film
Sealing Film Roll
UltraClear Sealing Film
Dimensions (L x W):
115 x 500 m
127.2 x 79.5 mm
133.3 x 79.2 mm
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
78 x 610 m
Not Available
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 100
Pack of 1000
Pack of 50
Pack of 500
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Product Type Dimensions (L x W) Sterility unitSize
11336254 Rayon Breathable Sealing Film 153 x 82.3 mm Sterile Pack of 50
12732796 Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile Pack of 1000
11326254 Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile Pack of 100
12742796 Rayon Breathable Sealing Film 153 x 82.3 mm Sterile Pack of 500
12623616 Aluminum Sealing Film Roll 115 x 500 m Non-sterile Each
12633726 Aluminum Sealing Film Roll 78 x 610 m Non-sterile Each
12613616 Polyester CyclerSeal Sealing Film Roll 115 x 500 m Non-sterile Each
12645926 Polyester CyclerSeal Sealing Film Roll 78 x 610 m Non-sterile Each
12557947 Litto or Ethylene or Vinyl Heat Seal Film 127.2 x 79.5 mm Non-sterile Pack of 100
12752796 Aluminum Sealing Film 146 x 79.2 mm Non-sterile Pack of 500
12577947 Aluminum Sealing Film 146 x 79.2 mm Non-sterile Pack of 100
12762796 Aluminum Sealing Film 133.3 x 79.2 mm Non-sterile Pack of 500
12782796 Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile Pack of 500
11380215 Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile Pack of 100
12772796 Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile Pack of 500
11336074 Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile Pack of 100
13131824 Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile Pack of 500
12014538 Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile Pack of 100
13191604 Polyolefin UltraClear Sealing Film Not Available Non-sterile Pack of 500
11304325 Polyolefin UltraClear Sealing Film Not Available Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 11336254 Supplier Axygen™ Supplier No. BF400S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Offer great economy for disposable ease.

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.

Breathable

  • Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
  • Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
Aluminum
  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200
CyclerSeal
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
AxySeal
  • Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
  • Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
  • Available in both nonsterile and sterile packaging

TRUSTED_SUSTAINABILITY

Specifications

Material Rayon
Sterility Sterile
Length (English) 6.02 in.
Length (Metric) 153 mm
No. per Case 500/Cs.
No. per Pack 50/Pk.
Dimensions (L x W) 153 x 82.3 mm
Width (English) 3.24 in.
Width (Metric) 82.3 mm
For Use With (Application) Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, allow effective gas exchange for cellular and bacterial cultivation, while preventing contamination
Product Type Breathable Sealing Film
Thickness 50 μm
Temperature Range (Metric) -20°C to +80°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.