missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ LightCycler 480 Compatible PCR Plates

Product Code. 12709628 Shop All Axygen Scientific Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 12709628

Brand: Axygen™ PCR384LC480CNF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to fit the Roche LightCycler™ 480

The new 480 plate offers another great solution for real-time PCR. Thin walls coupled with the 480 platform deliver real-time PCR results in less than 40 minutes. Every well of every plate undergoes an automated QC inspection
for microscopic holes, which gives you peace of mind when handling your valuable samples.

  • Optimized for the new plate-based Roche Light Cycler 480
  • Ideal for real-time PCR, white polypropylene magnifies signal intensity and prevents cross-talk between wells
  • Thin walls reduce cycle times
  • SBS standard dimensions
  • Black printing of alphanumerics for easily identifying wells and orienting plates
  • 100% automated QC for microscopic holes
  • Working volume: 3μL

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Format Standard
Array 16 x 24
Color Clear
For Use With (Equipment) For Roche light cycler 480
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 3 μL
Sterility Non-sterile
For Use With (Application) RT-PCR
Quantity 50/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.