missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ High Clarity Microplates†

Designed for use in sample collection, combinatorial chemistry, storage and HTS applications

Brand:  Axygen P-DW-12-HC

Code : A0

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12646207

  • £144.00 / Pack of 50


Estimated Shipment
07-04-2021
Add to basket

Description

Description

Made of virgin polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.

  • 1.2mL volume per well
  • Ideal for sample collection, storage, and HTS
  • ANSI/SBS footprint for automation use
  • High clarity design allows visual confirmation of sample from bottom
  • 19 round well low-profile design for reduced space requirements
  • Ultra-flat surface ensures proper sealing of heat film
  • Centrifugation to 4000 RCF
  • Diamond polished wells allow complete sample recovery

Model P-DW-12-HC-S: Pre-sterilized

Specifications

Specifications

Standard
Designed for use in sample collection, combinatorial chemistry, storage and HTS applications
Non-sterile
8 x 12
ANSI, SBS
Polypropylene
96
Videos
SDS
Documents