Learn More
Axygen™ GEN3™ Multi-Barrier Pipette Tips
Product Code. 12812283 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
100 to 1000 μL
20 μL
Unit Size:
Pack of 1000
Pack of 4800
Pack of 5000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12812283 100 to 1000 μL Pack of 5000
12882273 1 to 200 μL Pack of 4800
12802283 0.1 to 10 μL Pack of 4800
12822453 100 to 1000 μL Pack of 1000
12892273 20 μL Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12812283 Supplier Axygen™ Supplier No. GEN31000LRS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offers the ultimate in protection against cross contamination using tri-filter technology
The new Axygen™ brand GEN3 multi-barrier filter pipet tips offer the ultimate in protection against cross contamination using tri-filter technology. These tips will prevent liquid or aerosol from passing through and will lock on contact. GEN3 tips effectively separate liquid from the self-sealing barrier using an additional protective layer of conventional polyethylene filter material. In addition to the self-sealing properties, our GEN3 multi-barrier pipet tips feature our exclusive Maxymum Recovery™ technology which reduces the amount of reagent or sample adhering to the pipet tips. Offered in Racks, sterile, tips are nonpyrogenic and certified human gDNA and PCR inhibition free and DNase-/RNase-free.
  • Multi-barrier self-sealing filter prevents aerosol or liquid cross contamination
  • Certified human gDNA and PCR inhibition, RNase-/DNase-free
  • Nonpyrogenic
  • Proprietary filter design prevents possible PCR inhibition
  • Maxymum Recovery technology for ultra-low retention and high yield
  • Sterile, no need to autoclave
  • Certifications
    GEN3 multi-barrier tips are certified human gDNA and PCR inhibition free. Certified DNase-/RNase-free and nonpyrogenic.
    TRUSTED_SUSTAINABILITY
    Specifications
    Volume (Metric) 100 to 1000 μL
    Filtered Filtered
    Sterility Sterile
    Autoclavable Not Autoclavable
    Clean Claims DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free
    Format Rack
    Packaging 100 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
    Product Line GEN3
    Material Polyethylene