Learn More
Axygen™ Gel Documentation System Accessory, UV Transmitting Tray
Product Code. 16683004 Shop All Axygen Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Gel Documentation System (GD-1000)
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
16683004 Gel Documentation System (GD-1000) Each
1 options
Product Code. 16683004 Supplier Axygen™ Supplier No. GDGCS
  • ONLINE EXCLUSIVE PRICE
    £250.00
    £244.95 / Pack of 1
    Save £5.05
    2% Off
Estimated Shipment: 08-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Gel Documentation System (GD-1000)
Protects UV transilluminator when cutting out bands
TRUSTED_SUSTAINABILITY
Specifications
Product Type UV Transmitting Tray
For Use With (Equipment) Gel Documentation System (GD-1000)