Learn More
Axygen™ Gel Cutting Tips: 1.1 x 4 mm
Universal fit for research grade pipettes.
Brand:  Axygen™ TGL-1140-R
Code : A0
Additional Details : Weight : 6.35000kg
 View more versions of this product
Product Code. 12538017
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Axygen™ Gel Cutting Tips are designed to remove bands from agarose gel. These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 μL pipettors
  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Nonsterile
Specifications
Specifications
1000 μL
Clear
DNase, RNase Free
48 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
Polypropylene
Non-sterile
1.1 x 4 mm
Rack
Yes
2, 5μL