missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Caps for PCR Tubes

Product Code. 10240900 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
MaxyClear
MaxyClear, w/writing surface
Orange
Red
Violet
Yellow
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
Unit Size:
Pack of 100
Pack of 125
Pack of 1250
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Closure Type Product Type unitSize
10240900 Blue Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12607385 Clear Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
10557330 Clear Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12547987 Blue Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12841510 Assorted Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12537987 Assorted Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12871510 Green Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12557987 Green Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12567987 Orange Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12881510 Orange Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12801520 Violet Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12587987 Violet Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12891510 Red Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12577987 Red Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12597987 Yellow Domed Cap 8-Strip Domed PCR Tube Cap Pack of 125
12811520 Yellow Domed Cap 8-Strip Domed PCR Tube Cap Pack of 1250
12609535 MaxyClear, w/writing surface Flat Cap 8-Strip Flat PCR Tube Cap with Writing Surface Pack of 125
10666690 MaxyClear Flat Cap 8-Strip Flat PCR Tube Cap Pack of 1250
11320155 MaxyClear Flat Cap 8-Strip Flat PCR Tube Cap Pack of 125
15409073 Clear Domed Cap PCR Caps Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 10240900 Supplier Axygen™ Supplier No. PCR02CPB

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

8-Strip domed or flat strip caps for Axygen PCR tubes

Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Blue
Color Blue
Volume (Metric) 0.2 mL
Quantity 125/Pk.
Closure Material Polypropylene
For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Format Strip
Product Type 8-Strip Domed PCR Tube Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.