missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Caps for PCR Tubes

Product Code. 12577987 Shop All Axygen Scientific Products
Click to view available options
Color:
Assorted
Blue
Clear
Green
MaxyClear
MaxyClear, w/writing surface
Orange
Red
Violet
Yellow
Closure Type:
Domed Cap
Flat Cap
Product Type:
8-Strip Domed PCR Tube Cap
8-Strip Flat PCR Tube Cap
8-Strip Flat PCR Tube Cap with Writing Surface
PCR Caps
Unit Size:
Pack of 100
Pack of 125
Pack of 1250
This item is not returnable. View return policy

Product Code. 12577987

Brand: Axygen™ PCR02CPR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

8-Strip domed or flat strip caps for Axygen PCR tubes

Axygen polypropylene, thin wall PCR tubes with and without caps are designed to fit standard 96-well blocks and are available in an assortment of colors. The consistent and ultra-thin wall allows precise thermal transfer. An even insertion cap ensures a tight fit to reduce evaporation.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Red
Color Red
Volume (Metric) 0.2 mL
Quantity 125/Pk.
Closure Material Polypropylene
For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Format Strip
Product Type 8-Strip Domed PCR Tube Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.