Learn More
Axygen™ Axypet™ Pro Single Channel Pipettor
Product Code. 16362257 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.1 to 2 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
5 to 50 μL
Color:
Blue
Red
Yellow
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color unitSize
16362257 100 to 1000 μL Blue Each
16312267 0.1 to 2 μL Red Each
16342257 0.5 to 10 μL Red Each
16322267 2 to 20 μL Yellow Each
16342267 5 to 50 μL Yellow Each
16352257 10 to 100 μL Yellow Each
16332267 20 to 200 μL Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 16362257 Supplier Axygen™ Supplier No. AP1000P
Only null left
Add to basket
Add to basket
Corning™ Axygen™ Axypet™ Pro Pipettors are engineered to provide the highest levels of comfort, accuracy, and precision.
Axygen™ Axypet™ Pro Pipettors have been engineered to provide the highest levels of ergonomics, accuracy, and precision. Lightweight construction, contoured shape of the handle, and 4-digit counter were designed to ensure comfortable pipetting. All pipettors feature smooth plunger movement and extremely low pipetting forces to reduce the wrist strain and fatigue (RSI). Colored pushbuttons (included) can help define user or application for sample safety and lower risk of cross contamination.
  • Ergonomic handle design for perfect fit for both right and left hand
  • Convenient one-hand volume setting with auto-lock system prevents accidental volume change during work
  • 4-digit counter for greater precision, perfectly positioned for visibility at all times
  • Single-channel pipettors are compatible with very narrow tubes
  • Fully autoclavable and UV-resistant for sample protection against contaminations
  • The multi-channel shafts retract individually for perfect tip loading and easier tip ejection
  • Easy in-lab calibration helps you quickly adjust your pipettor to various liquid types
  • Individually tested and supplied with a certificate of quality
Learn more from North America
Learn more from Europe, Middle East, and Africa
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±2.5 to ±0.6
Certifications/Compliance ISO8655
Autoclavable Autoclavable
Color Blue
Grip Type Ergonomic
Number of Channels 1
Product Line Axypet Pro
Type Pipette
Volume (Metric) 100 to 1000 μL
Finger Style Ergonomic
Includes Instruction manual, QC Certificate, In-lab calibration tool
Increments 1 μL
Precision ≤0.6 to ≤0.2
Quantity 1
Warranty 3 months