missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Axypet-16 Pipette Tips

Product Code. 12792936 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 19200
Pack of 3840
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
12792936 Non-sterile Pack of 19200
11344245 Sterile Pack of 3840
12702946 Sterile Pack of 19200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12792936 Supplier Axygen™ Supplier No. T1536LR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For 1536-well pipetting

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention and high yield
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 30 μL
Sterility Non-sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) For 1536-well pipetting
Format Rack
Tip Style Low Retention
For Use With (Equipment) 1536-Well Pipetting
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.