Learn More
Axygen™ AxyMats™ Pierceable Sealing Mats for 24 Well Deep Well Plates
Minimize evaporation in wide range of temperatures in PCR and storage applications.
Brand:  Axygen™ AM-24-SQ-S
516.90 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.13000kg
 View more versions of this product
Product Code. 15551383
  • £723.00 / Pack of 50
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications
  • AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved
  • They can be reused following a bleach wash and ethanol rinse protocol.
  • Mfr. No. AM-24-SQ-S
Specifications
Specifications
Silicone
Autoclavable
5
Sealing Mat
Sterile
24
10
24-well Deep Well Plate with Square Wells