Learn More
Axygen™ 96-Well PCR Microplates
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications.
Brand: Axygen™ PCR-96-C-S
Code : A0
Additional Details : Weight : 1.50000kg
Description
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.
- 96-well PCR microplates
- Working volume is 300μL
- Available in a variety of colors
- Sold 10 plates/unit; 5 units/Cs.
Specifications
RNase, DNase-free | |
8 x 12 | |
Polypropylene | |
Non-sterile | |
Clear |
Standard | |
Clear | |
96 | |
PCR thermal transfer, RT-PCR applications |