missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Axygen™ 96 well segmented PCR microplate is designed to fit standard 96 well blocks. Plate is pre-scored to break apart into four 24 well sections. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.
RNase-/DNase-free Nonpyrogenic Nonsterile Manufactured with 99.9% pure virgin polypropylene 96-well Segmented PCR Plate Application/Compatibility: Standard thermal cycling, Beckman CEQ Sequencers
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | RNase, DNase-free |
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Clear |
| For Use With (Equipment) | Beckman CEQ Sequencers |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
