Learn More
- Home
- Products
- Microplates
- Nucleic Acid Reaction Assay Microplates
- Axygen™ 96-well PCR Microplates
Axygen™ 96-well PCR Microplates
Product Code. 11370185 Shop All Axygen Scientific Products| Product Code. | Color | Skirt Style | Format | unitSize |
|---|---|---|---|---|
| 11370185 | Black | Full | Standard | Pack of 10 |
| 12799438 | Clear | Elevated | Standard | Pack of 50 |
| 11396124 | Clear | Elevated | Standard | Pack of 10 |
| 12112580 | Clear | Unskirted | Standard | Pack of 50 |
| 11391804 | Clear | Unskirted | Standard | Pack of 10 |
| 11330185 | Clear | Unskirted | Flat Top | Pack of 25 |
| 12335193 | Clear | Unskirted | Flat Top | Pack of 100 |
| 12719458 | Clear | Full | Standard | Pack of 50 |
| 11380185 | Clear | Full | Standard | Pack of 10 |
| 11320185 | Black | Unskirted | Flat Top | Pack of 25 |
| 12799448 | Blue | Full | Standard | Pack of 50 |
| 11360185 | Blue | Full | Standard | Pack of 10 |
| 12779458 | Yellow | Full | Standard | Pack of 50 |
| 11340195 | Yellow | Full | Standard | Pack of 10 |
| 11300195 | Orange | Full | Standard | Pack of 10 |
| 12739458 | Orange | Full | Standard | Pack of 50 |
| 12779448 | White | Unskirted | Flat Top | Pack of 100 |
| 12749458 | Red | Full | Standard | Pack of 50 |
| 11310195 | Red | Full | Standard | Pack of 10 |
| 12709458 | Black | Full | Standard | Pack of 50 |
| 12789448 | Assorted | Full | Standard | Pack of 50 |
| 11350185 | Assorted | Full | Standard | Pack of 10 |
| 11320205 | Clear | Unskirted | Elevated Wells | Pack of 10 |
| 12799478 | Clear | Unskirted | Elevated Wells | Pack of 50 |
| 12567787 | Clear | Half | Standard | Pack of 10 |
| 10332390 | Clear | Half | Standard | Pack of 50 |
| 13164824 | Clear | Half | Single Notch Corner | Pack of 50 |
| 12537797 | Clear | Half | Single Notch Corner | Pack of 10 |
| 12769458 | White | Full | Standard | Pack of 50 |
| 11330195 | White | Full | Standard | Pack of 10 |
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | RNase-, DNase-free |
| Format | Standard |
| Array | 8 x 12 |
| Color | Black |
| Product Type | Flat top |
| For Use With (Equipment) | Standard 96 well blocks |
| Skirt Style | Full |
| Material | Polypropylene |
| Volume (Metric) Well | 200 μL |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Round |
| For Use With (Application) | PCR thermal transfer, RT-PCR applications |
| Show More Show Less |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.