Learn More
Axygen™ 4- Strip PCR Tubes and Caps for Corbett Rotor-Gene™
Easy snap and lock caps
Brand: Axygen™ PCR-0104-C
237.37 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Code : A0
Additional Details : Weight : 0.13500kg
Description
Axygen™ 0.1mL PCR tubes are specifically designed for use with the Corbett Rotorgene system. There are 4 tubes per strip and 4 caps per strip.
- Certified RNase-/DNase-free
- Nonpyrogenic
- Nonsterile
- Manufactured with 99.9% pure virgin polypropylene
Order Info
Tubes and caps are in strips of four and are in separate bags for ease of handling and labeling.
Specifications
RNase, DNase and non-pyrogenic free | |
Clear | |
250/Pk. | |
Corbett™ Rotor-Gene™ system | |
Non-sterile | |
Snap and Lock Plug Seal Flat Cap | |
4-Strip PCR Tube And Flat Cap |
Clear | |
0.1 mL | |
Polypropylene | |
Conical | |
Polypropylene | |
Strip |