missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 4- Strip PCR Tubes and Caps for Corbett Rotor-Gene™
Shop All Axygen Scientific Products

Click to view available options
Color:
Clear
Unit Size:
Pack of 2500
Description
Axygen™ 0.1mL PCR tubes are specifically designed for use with the Corbett Rotorgene system. There are 4 tubes per strip and 4 caps per strip.
- Certified RNase-/DNase-free
- Nonpyrogenic
- Nonsterile
- Manufactured with 99.9% pure virgin polypropylene
Order Info
Tubes and caps are in strips of four and are in separate bags for ease of handling and labeling.
Specifications
Specifications
| Certifications/Compliance | RNase, DNase and non-pyrogenic free |
| Closure Color | Clear |
| Color | Clear |
| Volume (Metric) | 0.1 mL |
| Quantity | 250/Pk. |
| Closure Material | Polypropylene |
| For Use With (Equipment) | Corbett™ Rotor-Gene™ system |
| Shape | Conical |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction