missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 384-well Skirted PCR Microplates

Product Code. 11350155 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Orange
Red
Violet
White
Yellow
Includes:
Double notch
Single notch
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 50
Volume (Metric) Well:
40 μL
50 μL
55 μL
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Includes unitSize Volume (Metric) Well
11350155 Blue Double notch Pack of 10 -
12719418 Blue Double notch Pack of 50 -
11380155 Clear Single notch Pack of 10 50 μL
12749418 Clear Single notch Pack of 50 50 μL
12759418 Green Double notch Pack of 50 -
11390155 Green Double notch Pack of 10 -
11300165 Orange Double notch Pack of 10 -
12709438 Orange Double notch Pack of 50 -
12516887 White Single notch Pack of 10 55 μL
10068469 White Single notch Pack of 50 55 μL
12739438 Clear Double notch Pack of 100 50 μL
12567817 Clear Double notch Pack of 20 50 μL
11310165 Red Double notch Pack of 10 40 μL
12719438 Red Double notch Pack of 50 40 μL
11350165 Violet Double notch Pack of 10 40 μL
12759438 Violet Double notch Pack of 50 40 μL
12769438 Yellow Double notch Pack of 50 -
11360165 Yellow Double notch Pack of 10 -
15328301 Clear Single notch Pack of 50 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 11350155 Supplier Axygen™ Supplier No. PCR384B

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Single and double notch 384-well PCR plates

Sealing Options: Axymats, heat sealing, adhesive films and foils.

  • Available in 50μL and 55μL
  • Single and double notch corner

Skirted (50μL)

  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.

Colors available by special order.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance RNase, DNase-free
Format Standard
Array 16 x 24
Color Blue
For Use With (Equipment) N
Material Polypropylene
No. of Wells 384
Volume (Metric) Working 40 μL
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR, thermal transfer applications
Includes Double notch
Well Color Blue
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.