missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 384-well Skirted PCR Microplates

Product Code. 11380155 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Orange
Red
Violet
White
Yellow
Volume (Metric) Well:
40 μL
50 μL
55 μL
Includes:
Double notch
Single notch
Unit Size:
Pack of 10
Pack of 100
Pack of 20
Pack of 50
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Well Includes unitSize
11380155 Clear 50 μL Single notch Pack of 10
12719418 Blue - Double notch Pack of 50
11350155 Blue - Double notch Pack of 10
12749418 Clear 50 μL Single notch Pack of 50
12759418 Green - Double notch Pack of 50
11390155 Green - Double notch Pack of 10
11300165 Orange - Double notch Pack of 10
12709438 Orange - Double notch Pack of 50
12516887 White 55 μL Single notch Pack of 10
10068469 White 55 μL Single notch Pack of 50
12739438 Clear 50 μL Double notch Pack of 100
12567817 Clear 50 μL Double notch Pack of 20
11310165 Red 40 μL Double notch Pack of 10
12719438 Red 40 μL Double notch Pack of 50
11350165 Violet 40 μL Double notch Pack of 10
12759438 Violet 40 μL Double notch Pack of 50
12769438 Yellow - Double notch Pack of 50
11360165 Yellow - Double notch Pack of 10
15328301 Clear 50 μL Single notch Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
This item is not returnable. View return policy
Product Code. 11380155 Supplier Axygen™ Supplier No. PCR384CS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Single and double notch 384-well PCR plates

Sealing Options: Axymats, heat sealing, adhesive films and foils.

  • Available in 50μL and 55μL
  • Single and double notch corner

Skirted (50μL)

  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.

Colors available by special order.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance RNase, DNase-free
Format Standard
Array 16 x 24
Color Clear
Skirt Style Full
Material Polypropylene
Volume (Metric) Well 50 μL
No. of Wells 384
Volume (Metric) Working 40 μL
Sterility Sterile
Well Shape Round
For Use With (Application) PCR
Includes Single notch
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.