missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Axygen™ 384 well skirted PCR microplate, clear, is designed to fit standard 384 well blocks. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Automation friendly full skirt design can be sealed with Axymat or sealing film.
RNase-/DNase-free Nonpyrogenic Nonsterile Manufactured with 99.9% pure virgin polypropylene 384-well double notch PCR plate Robotic friendly full skirt design Application/Compatibility: Standard thermal cycling
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | RNase, DNase-free |
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 384 |
| Volume (Metric) Working | 40 μL |
| Sterility | Non-sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?