missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 384-well Skirted PCR Microplates

Single and double notch 384-well PCR plates

Brand:  Axygen™ PCR-384-RGD-C

Code : 68

Additional Details : Weight : 3.18000kg

 View more versions of this product

Product Code. 12739438

  • £860.00 / Pack of 100
Estimated Shipment: 11-09-2024
to see stock.
Add to basket

Description

Description

Sealing Options: Axymats, heat sealing, adhesive films and foils.

  • Available in 50μL and 55μL
  • Single and double notch corner

Skirted (50μL)

  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.

Colors available by special order.

Specifications

Specifications

No Barcode
Standard
16 x 24
Full
50 μL
49.5 μL
Round
Double notch
20/Pk.
RNase, DNase-free
Without Lid
Clear
Polypropylene
384
Non-sterile
For standard thermal cycling, ABI sequencing, automation applications
100/Cs.
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers