missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Maxymum Recovery™ 300 μL Universal Pipetter Tips

Product Code. 11374255 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 10000
Pack of 4800
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format unitSize
11374255 Sterile Rack Pack of 960
12541348 Non-sterile Bulk Pack of 10000
11364255 Non-sterile Bulk Pack of 1000
12699595 Non-sterile Rack Pack of 960
12716609 Non-sterile Rack Pack of 4800
12726609 Sterile Rack Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11374255 Supplier Axygen™ Supplier No. T350CLRS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Maxymum Recovery tips fit popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Fine-Point
  • Maxymum Recovery technology for ultra-low retention and high yield

Order Info

  • Bulk: 1000 tips/pack, 240 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 300 μL
Sterility Sterile
Color Clear
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Rack
Tip Style Low Retention, Universal
Packaging Hinged rack
For Use With (Equipment) Universal Fit
Product Line Maxymum Recovery
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.