missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 24- and 32-Well PCR Microplates, Clear, Nonsterile
GREENER_CHOICE

Product Code. 12799408 Shop All Axygen Scientific Products
Change view
Click to view available options
No. of Wells:
24
32
Unit Size:
Pack of 10
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells unitSize
12799408 32 Pack of 50
11310155 24 Pack of 10
12789408 24 Pack of 50
11330155 32 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12799408 Supplier Axygen™ Supplier No. PCR32C
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to fit a wide variety of thermal cyclers

  • Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Lid Without Lid
Color Clear
Product Type PCR Microplate, without Lid
Material Polypropylene
No. of Wells 32
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.