missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 1250μL Universal Pipette Tips

Product Code. 11326334 Shop All Axygen Scientific Products
Change view
Click to view available options
Packaging:
Bulk
Rack
Unit Size:
Pack of 1152
Pack of 3456
Pack of 5000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
11326334 Rack Pack of 1152
13107664 Rack Pack of 3456
12531308 Bulk Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326334 Supplier Axygen™ Supplier No. MTX1250CR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Matrix compatible tips in clear color

  • Matrix-style tips fits Matrix Impact™ and Impact2™ brand pipettors
  • Maxymum Recovery™ tips allows complete retrieval of precious samples
  • Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 15 to 1250μL
  • Available in bulk or racked formats

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1250 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Certifications/Compliance RNase, DNase-free
Packaging Rack
For Use With (Equipment) Thermo Scientific Matrix Impact
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.