missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Maxymum Recovery™ 20 μL Ultra Micro Tips

Product Code. 12653706 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 20000
Pack of 4800
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format unitSize
12653706 Sterile Rack Pack of 960
12551368 Non-sterile Bulk Pack of 20000
12796609 Non-sterile Rack Pack of 4800
12611656 Non-sterile Rack Pack of 960
12706619 Sterile Rack Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12653706 Supplier Axygen™ Supplier No. T400LRS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are RNase-/DNase-free and nonpyrogenic. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic andRNase-/DNase-free.
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention and high yield
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 20 μL
Sterility Sterile
Color Clear
Clean Claims DNase, RNase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Micro Point, Low Retention
Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment) Eppendorf, Gilson Pipetman P2, Gilson Pipetman P10
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.