Learn More
Axygen™ 10μL Microvolume Pipette Tips, Non-Filtered
Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
Brand: Axygen™ TXL-10-R-S
This item is not returnable.
View return policy
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are certified to be RNase-/DNase-free and nonpyrogenic. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- Non-pyrogenic
- Microvolume
- Non-filtered
- Maxymum Recovery technology for ultra-low retention
Specifications
0.1 to 10 μL | |
Sterile | |
USP 3127 | |
Allows recovery of costly samples with improved precision and accuracy | |
Extended Length, Microvolume | |
Gilson™ Pipetman™ P2, Gilson™ Pipetman™ P10 |
Non-filtered | |
Clear | |
DNase, Rnase Free | |
Rack | |
96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs | |
Polypropylene |