missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 10μL Microvolume Pipette Tips, Non-Filtered

Product Code. 11364315 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
Unit Size:
Pack of 960
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11364315 0.1 to 10 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11364315 Supplier Axygen™ Supplier No. TXL10RS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are certified to be RNase-/DNase-free and nonpyrogenic. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • Non-pyrogenic
  • Microvolume
  • Non-filtered
  • Maxymum Recovery technology for ultra-low retention
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Certifications/Compliance USP 3127
Clean Claims DNase, Rnase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Extended Length, Microvolume
Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
For Use With (Equipment) Gilson Pipetman P2, Gilson Pipetman P10
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.