Learn More
Axygen™ 1.5 mL Self-Standing Screw Cap Tubes
Product Code. 11330395 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Assorted
Brown
Clear
Gray
Green
Orange
Pink
Red
Violet
White
Yellow
Unit Size:
Pack of 4000
Pack of 500
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Closure Color unitSize
11330395 Non-sterile Red Pack of 500
11340385 Non-sterile Assorted Pack of 500
12779538 Sterile Assorted Pack of 4000
11350385 Sterile Assorted Pack of 500
11366044 Non-sterile Clear Pack of 500
13107464 Non-sterile Clear Pack of 4000
11380385 Sterile Clear Pack of 500
12719548 Sterile Clear Pack of 4000
11370385 Non-sterile Brown Pack of 500
12799538 Non-sterile Brown Pack of 4000
13124624 Non-sterile Green Pack of 4000
11300395 Non-sterile Gray Pack of 500
12739548 Non-sterile Gray Pack of 4000
12789548 Non-sterile Orange Pack of 4000
11310395 Non-sterile Orange Pack of 500
12799548 Non-sterile Pink Pack of 4000
11320395 Non-sterile Pink Pack of 500
12709558 Non-sterile Red Pack of 4000
12719558 Non-sterile Violet Pack of 4000
11340395 Non-sterile Violet Pack of 500
12729558 Non-sterile White Pack of 4000
11354165 Non-sterile White Pack of 500
12749558 Non-sterile Yellow Pack of 4000
11374165 Non-sterile Yellow Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Product Code. 11330395 Supplier Axygen™ Supplier No. SCT150SSR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Features O-ring manufactured from material that is resistant to many commonly-used laboratory chemicals
For collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Pre-sterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Red
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Material Polypropylene
Format Non-looped
Includes O-ring
Sterility Non-sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g