Learn More
Axygen™ 1.5 mL Conical Screw Cap Tubes
Product Code. 11360375 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Assorted
Blue
Brown
Clear
Gray
Green
Orange
Pink
Red
Violet
White
Yellow
Unit Size:
Pack of 4000
Pack of 500
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Closure Color unitSize
11360375 Non-sterile Brown Pack of 500
12769528 Non-sterile Brown Pack of 4000
11330375 Non-sterile Assorted Pack of 500
12739528 Non-sterile Assorted Pack of 4000
13181774 Sterile Assorted Pack of 4000
11340375 Sterile Assorted Pack of 500
11350375 Non-sterile Blue Pack of 500
11370375 Non-sterile Clear Pack of 500
12176830 Non-sterile Clear Pack of 4000
12186830 Sterile Clear Pack of 4000
11380375 Sterile Clear Pack of 500
12799528 Non-sterile Green Pack of 4000
11300385 Non-sterile Gray Pack of 500
12709538 Non-sterile Gray Pack of 4000
12739538 Non-sterile Orange Pack of 4000
12749538 Non-sterile Pink Pack of 4000
11320385 Non-sterile Pink Pack of 500
12759538 Non-sterile Red Pack of 4000
11330385 Non-sterile Red Pack of 500
12759558 Non-sterile Violet Pack of 4000
11384165 Non-sterile Violet Pack of 500
12769558 Non-sterile White Pack of 4000
11394165 Non-sterile White Pack of 500
11304175 Non-sterile Yellow Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Product Code. 11360375 Supplier Axygen™ Supplier No. SCT150BR
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
With chemical-resistant O-ring
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order
  • Caps available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Brown
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Material Polypropylene
Format Non-looped
Includes O-ring
Sterility Non-sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g