Learn More
Axygen™ 0.5 mL Self Standing Screw Cap Tubes
Product Code. 11340365 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Closure Color:
Assorted
Blue
Brown
Clear
Gray
Green
Orange
Red
Violet
White
Yellow
Unit Size:
Pack of 4000
Pack of 500
37 product options available for selection
Product selection table with 37 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Closure Color unitSize
11340365 Non-sterile Violet Pack of 500
13194624 Non-sterile Assorted Pack of 4000
11330355 Non-sterile Assorted Pack of 500
12779498 Sterile Assorted Pack of 4000
11340355 Sterile Assorted Pack of 500
11350355 Non-sterile Blue Pack of 500
13167394 Non-sterile Blue Pack of 4000
13151784 Non-sterile Clear Pack of 4000
11370355 Non-sterile Clear Pack of 500
12709508 Sterile Brown Pack of 4000
12599007 Sterile Brown Pack of 500
12799498 Non-sterile Brown Pack of 4000
11360355 Non-sterile Brown Pack of 500
12739508 Sterile Clear Pack of 4000
11380355 Sterile Clear Pack of 500
13161784 Non-sterile Green Pack of 4000
11390355 Non-sterile Green Pack of 500
12759508 Non-sterile Gray Pack of 4000
11300365 Non-sterile Gray Pack of 500
12559017 Sterile Gray Pack of 500
10567270 Sterile Gray Pack of 4000
10337839 Sterile Green Pack of 4000
12569017 Sterile Green Pack of 500
11310365 Non-sterile Orange Pack of 500
12719518 Non-sterile Red Pack of 4000
11330365 Non-sterile Red Pack of 500
10312250 Sterile Red Pack of 4000
12519027 Sterile Red Pack of 500
12539027 Sterile Violet Pack of 500
12749518 Sterile Violet Pack of 4000
12739518 Non-sterile Violet Pack of 4000
12759518 Non-sterile White Pack of 4000
11350365 Non-sterile White Pack of 500
12589027 Sterile Yellow Pack of 500
10713392 Sterile Yellow Pack of 4000
12789518 Non-sterile Yellow Pack of 4000
11370365 Non-sterile Yellow Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 37 options available.
37 options
Product Code. 11340365 Supplier Axygen™ Supplier No. SCT050SSV
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Features O-ring manufactured for resistant to many commonly-used laboratory chemicals
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile tubes
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Wide range of colored tubes for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Violet
Closure Material Polypropylene
Capacity (Metric) 0.5 mL
Material Polypropylene
Format Non-looped
Includes O-ring
Sterility Non-sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g