Learn More
Axygen™ 0.5 mL Conical Screw Cap Tubes: Presterilized
Product Code. 11370345 Shop All Axygen Scientific Products
Change view
Click to view available options
Closure Color:
Assorted
Clear
Unit Size:
Pack of 4000
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color unitSize
11370345 Clear Pack of 500
12769488 Assorted Pack of 4000
11330345 Assorted Pack of 500
12709498 Clear Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11370345 Supplier Axygen™ Supplier No. SCT050CS
Only null left
Add to basket
Add to basket
Features chemical resistant O-ring
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping of restriction enzymes and other reagents.
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Available in range of colors for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Certifications/Compliance RNase, DNase, pyrogen-free
Closure Color Clear
Closure Material Polypropylene
Capacity (Metric) 0.5 mL
Color Clear
Material Polypropylene
Format Non-looped
Includes O-ring
No. per Pack 500/Pk.
Packaging 500 Tubes and caps/Pk.; 8 Packs/Cs.
Sterility Sterile
Graduated No
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g