missing translation for 'onlineSavingsMsg'
Learn More

AXUD1 Antibody (5E8), Novus Biologicals™

Product Code. 18423879 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423879 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423879 Supplier Novus Biologicals Supplier No. H00064651M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AXUD1 Monoclonal antibody specifically detects AXUD1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen AXUD1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5E8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH38949
Gene Alias AXIN1 up-regulated 1, Axin-1 up-regulated gene 1 protein, AXUD1DKFZp566F164, CSRNP-1, cysteine/serine-rich nuclear protein 1, cysteine-serine-rich nuclear protein 1, FAM130B, Protein URAX1, TAIP3, TAIP-3TGF-beta-induced apoptosis protein 3, TGF-beta induced apoptosis protein 3, URAX1
Host Species Mouse
Immunogen AXUD1 (AAH38949, 1 a.a. ∽ 589 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTGLLKRKFDQLDEDNSSVSSSSSSSGCQSRSCSPSSSVSRAWDSEEEGPWDQMPLPDRDFCGPRSFTPLSILKRARRERPGRVAFDGITVFYFPRCQGFTSVPSRGGCTLGMALRHSACRRFSLAEFAQEQARARHEKLRQRLKEEKLEMLQWKLSAAGVPQAEAGLPPVVDAIDDASVEEDLAVAVAGGRLEEVSFLQPYPARRRRALLRASGVRRIDREEKRELQALRQSREDCGCHCDRICDPETCSCSLAGIKCQMDHTAFPCGCCREGCENPMGRVEFNQARVQTHFIHTLTRLQLEQEAESFRELEAPAQGSPPSPGEEALVPTFPLAKPPMNNELGDNSCSSDMTDSSTASSSASGTSEAPDCPTHPGLPGPGFQPGVDDDSLARILSFSDSDFGGEEEEEEEGSVGNLDNLSCFHPADIFGTSDPGGLASWTHSYSGCSFTSGILDENANLDASCFLNGGLEGSREGSLPGTSVPPSMDAGRSSSVDLSLSSCDSFELLQALPDYSLGPHYTSQKVSDSLDNIEAPHFPLPGLSPPGDASSCFLESLMGFSEPAAEALDPFIDSQFEDTVPASLMEPVPV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 64651
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.