missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Axl Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EMAXLX5

Additional Details : Weight : 0.33800kg

 View more versions of this product

Product Code. 17195679

  • £2320.00 / Each
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

The Mouse Axl ELISA quantitates Ms Axl in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Axl. Principle of the method The Mouse Axl solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

AXL (also known as tyrosine-protein kinase receptor UFO) is a cell surface receptor. The AXL receptor transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6 (GAS6). It is involved in the stimulation of cell proliferation. This receptor can also mediate cell aggregation by homophilic binding. AXL is an essential epithelial-to-mesenchymal transition-induced regulator of breast cancer metastasis.
Specifications

Specifications

Q00993
1 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
26362
<12%
Assay Diluent Concentrate, Biotinylated Detection Antibody, Wash Buffer, Standard, Adhesive Plate Covers, Stop Solution, Chromogen, Pre-coated 96 well plates, SAV-HRP
5 x 96 Tests
Inflammation
2°C to 8°C
Mouse
4 hr. 45 min.
1.0 pg/mL to 250 pg/mL
1 pg/mL to 250 pg/mL
Biotin
Proteases, Peptidases & Inhibitors
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL- 4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
Mouse
Colorimetric Microplate Reader
AI323647,Ark,Tyro7,Ufo
<10%
HRP
RUO
Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L
AI323647, Ark, Tyro7, Ufo
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers