missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Axl Monoclonal Antibody (DS7HAXL), Brilliant Violet™ 421, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£153.00 - £321.00
Specifications
| Antigen | Axl |
|---|---|
| Clone | DS7HAXL |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089220
|
Invitrogen™
404-1087-41 |
25 Tests |
£153.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30087538
![]() |
Invitrogen™
404-1087-42 |
100 Tests |
£321.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Axl Monoclonal antibody specifically detects Axl in Human samples. It is validated for Flow CytometrySpecifications
| Axl | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| DS7HAXL | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Human | |
| P30530 | |
| 558 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| AXL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title