missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AXL Human ELISA Kit

Product Code. 15994204
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 15994204

Brand: Invitrogen™ BMS2286

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sandwich ELISA Kit

The Human AXL ELISA quantitates Hu AXL in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu AXL.

Principle of the method

The Human AXL solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

AXL (also known as tyrosine-protein kinase receptor UFO) is a cell surface receptor. The AXL receptor transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6 (GAS6). It is involved in the stimulation of cell proliferation. This receptor can also mediate cell aggregation by homophilic binding. AXL is an essential epithelial-to-mesenchymal transition-induced regulator of breast cancer metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P30530
Assay Range 31.25-2000 pg/ml
Assay Sensitivity 7.457 pg/ml
Conjugate Biotin
Product Type ELISA Kit
Sample Type Cell Lysates, Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 558
Gene Symbol AXL
Interassay CV 10%
Intraassay CV 8.6%
Kit Contents Pre-coated 96-well Plate, Standard (2 vials), Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromagen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Serum, 10 μL; Plasma, 10 μL; Cell Lysate,50 μL; Supernatant, 50 μL
Storage Requirements 2°C to 8°C
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.