missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Axl Monoclonal antibody specifically detects Axl in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface), CyTOF
Specifications
Specifications
| Applications | Flow Cytometry (Cell Surface), CyTOF |
| Classification | Monoclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL |
| Host Species | Mouse |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?