missing translation for 'onlineSavingsMsg'
Learn More

Axitinib, MedChemExpress

Product Code. 16454406
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16454406

Brand: Medchem Express HY10065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Axitinib is a multi-targeted tyrosine kinase inhibitor with IC50s of 0.1, 0.2, 0.1-0.3, 1.6 nM for VEGFR1, VEGFR2, VEGFR3 and PDGFRβ, respectively.

Axitinib is a multi-targeted tyrosine kinase inhibitor with IC50s of 0.1, 0.2, 0.1-0.3, 1.6 nM for VEGFR1, VEGFR2, VEGFR3 and PDGFRβ, respectively.

Specifications

Chemical Name or Material Axitinib
Physical Form Solid
CAS 319460-85-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H18N4OS
For Use With (Application) Cancer-Kinase/protease
Synonym AG-013736
Solubility Information DMSO : 20.83 mg/mL (53.90 mM; Need ultrasonic)
SMILES O=C(C1=C(SC2=CC3=C(C(/C=C/C4=CC=CC=N4)=NN3)C=C2)C=CC=C1)NC
Molecular Weight (g/mol) 386.47
Formula Weight 386.47
Percent Purity 98.21%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.