missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axitinib, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Description
Axitinib is a multi-targeted tyrosine kinase inhibitor with IC50s of 0.1, 0.2, 0.1-0.3, 1.6 nM for VEGFR1, VEGFR2, VEGFR3 and PDGFRβ, respectively.
Specifications
Specifications
| Chemical Name or Material | Axitinib |
| Physical Form | Solid |
| CAS | 319460-85-0 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18N4OS |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | AG-013736 |
| Solubility Information | DMSO : 20.83 mg/mL (53.90 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction