missing translation for 'onlineSavingsMsg'
Learn More

Axitinib Iodo Tetrahydropyran, TRC

Product Code. 30390691
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390691 1 mg 1mg
30402164 10 mg 10mg
30391248 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390691 Supplier Toronto Research Chemicals Supplier No. TRCI7194601MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Axitinib Iodo Tetrahydropyran
Analyte or Component Names Axitinib Iodo Tetrahydropyran
Shipping Condition Room Temperature
Molecular Formula C19 H20 I N3 O
Synonym 6-Iodo-3-(2-(pyridin-2-yl)ethyl)-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
InChI Formula InChI=1S/C19H20IN3O/c20-14-7-9-16-17(10-8-15-5-1-3-11-21-15)22-23(18(16)13-14)19-6-2-4-12-24-19/h1,3,5,7,9,11,13,19H,2,4,6,8,10,12H2
Solution Type Neat
SMILES Ic1ccc2c(CCc3ccccn3)nn(C4CCCCO4)c2c1
IUPAC Name 6-iodo-1-(oxan-2-yl)-3-(2-pyridin-2-ylethyl)indazole
Industry Type Pharmaceutical
Molecular Weight (g/mol) 433.286
Quantity 1 mg
Formula Weight 433.065
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.