missing translation for 'onlineSavingsMsg'
Learn More

Axitinib-d3, TRC

Product Code. 17949249
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
10 mg
Unit Size:
0.25mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949249 10 mg 10mg
17908229 0.25 mg 0.25mg
17928329 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17949249 Supplier Toronto Research Chemicals Supplier No. TRCA79465210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126623-89-9
Molecular Formula C22 D3 H15 N4 O S
Molecular Weight (g/mol) 389.488
Synonym Benzamide, N-(methyl-d3)-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1H-indazol-6-yl]thio]- (ACI), N-(Methyl-d3)-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1H-indazol-6-yl]thio]benzamide (ACI), 2-[[3-[(E)-2-Pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]-N-(trideuteriomethyl)benzamide, N-(Methyl-d3)-2-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]benzamide, Axitinib-d3, Axitinib-D3 (N-methyl-D3)
IUPAC Name 2-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]-N-(trideuteriomethyl)benzamide
SMILES [2H]C([2H])([2H])NC(=O)c1ccccc1Sc2ccc3c(\C=C\c4ccccn4)n[nH]c3c2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H18N4OS/c1-23-22(27)18-7-2-3-8-21(18)28-16-10-11-17-19(25-26-20(17)14-16)12-9-15-6-4-5-13-24-15/h2-14H,1H3,(H,23,27)(H,25,26)/b12-9+/i1D3
Formula Weight 389.139
Chemical Name or Material Axitinib-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.