missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 226.0°C to 228.0°C |
| Quantity | 250 mg |
| Solubility Information | Solubility aq. soln.: .. Other solubilities: soluble in methanol, dimethylsulfoxide |
| Formula Weight | 386.47 |
| Chemical Name or Material | Axitinib |
Safety and Handling
Product Identifier
- Axitinib
- Warning
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- H302-Harmful if swallowed.
- H400-Very toxic to aquatic life.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: N-Methyl-2-({ 3-[(E)-2-pyridin-2-ylvinyl]-1H-indazol-6-yl} thio)benzamide
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?