missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Axiom™ SwiftArrayStudio™ Consumables Kit

Product Code. 31782066 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
96 Reactions
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31782066 96 Reactions 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31782066 Supplier Applied Biosystems™ Supplier No. 952674
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SwiftArrayStudio Consumable Kit includes essential consumables for reliable operation of the SwiftArrayStudio instrument. Designed for consistent handling and workflow efficiency, it helps labs maintain throughput and performance while minimizing setup time and variability.

The Axiom SwiftArrayStudio Consumables Kit includes all SwiftArrayStudio instrument consumables required to process one Axiom SwiftArray 96F plate.

TRUSTED_SUSTAINABILITY

Specifications

Type Microarray Genotyping
Array Genotyping, Human Gene Expression, Carrier Screening Research, Copy Number, Cytogenetics
Number of Arrays 1 array
Format Array Plate
Species Human, Ag
No. of Samples 96 Reactions
Quantity 96 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.