missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Axiom™ SwiftArrayStudio™ Buffer Kit

Product Code. 31783843 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2x96 Arrays
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31783843 2x96 Arrays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31783843 Supplier Applied Biosystems™ Supplier No. 952638
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SwiftArrayStudio Buffer Kit provides all necessary reagents for two complete runs on the SwiftArrayStudio instrument. Designed for reliable performance and easy setup, it ensures consistent buffer quality and workflow efficiency, helping labs maintain throughput and data integrity with every use.

The Axiom SwiftArrayStudio Buffer Kit contains sufficient wash and buffer to process two 96F SwiftArrays.

Note: This buffer kit is included in the PangenomePro and PharmacoPro kits.

TRUSTED_SUSTAINABILITY

Specifications

Type Microarray Genotyping
Array Genotyping, DNA, Copy Number
Number of Arrays 2 arrays
Format Kit
Species Human, Ag
Includes Buffer Kit Only, No Arrays Included
No. of Samples 2x96 Arrays
Quantity 2x96 Arrays

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.