missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Axiom™ PangenomePro™ SwiftArray™ Kit

Product Code. 31782802 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Samples:
1x96F
2x96F
Unit Size:
96 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples unitSize
31782802 1x96F 96 reactions
31783275 2x96F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31782802 Supplier Applied Biosystems™ Supplier No. 952643
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PangenomePro delivers comprehensive, population-diverse genome-wide coverage in a high-throughput array format. Designed for inclusive GWAS and imputation studies, PangenomePro helps researchers generate more representative genetic insights across global populations.

The Axiom PangenomePro Array boasts the largest ethnic diversity coverage within the Axiom portfolio and enables target SNP identification, copy number detection and variant analysis, and HLA typing in a cost-effective assay with plug and play data analysis. It delivers ethnically diverse, genome-wide coverage in a high-throughput 96-array plate format, designed to support whole-genome imputation across global populations and help researchers build more representative genomic datasets. The array can serve as a core research tool for human genomics workflows including GWAS, population health initiatives, polygenic risk scores, pharmacogenomics, and drug discovery.

Features of the Axiom PangenomePro Array include:

  • Marker selection that emphasizes broad imputation power across European, African, admixed American, East Asian, and South Asian populations
  • Whole-genome analysis from as little as 100 ng genomic DNA with support for target SNP identification, CNV analysis, HLA and blood typing, and 'plug and play' data analysis workflows
  • GWAS imputation module (~850,000 markers) across ancestral populations
  • Evidence-based clinical/research markers that include references to public resources such as ACMG 73, ClinVar, NHGRI-GWAS catalog
  • Pharmacogenomics coverage aligned to guideline and knowledgebase citations (e.g., CPIC, PharmGKB, PharmaADME)
  • CNV analysis for fixed regions and de novo discovery across the genome
TRUSTED_SUSTAINABILITY

Specifications

Type Human Microarray Genotyping
Array Genotyping, DNA, Copy Number
Number of Arrays 1 array
Format 96-array Plate
Species Human
No. of Samples 1x96F
Quantity 96 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.