missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Axiom™ PangenomePro™ Plus SwiftArray™ Kit

Product Code. 31784113 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Samples:
1x96F
2x96F
Unit Size:
96 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples Unit Size
31784113 1x96F 96 reactions
31782384 2x96F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31784113 Supplier Applied Biosystems™ Supplier No. 952644
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PangenomePro delivers comprehensive, population-diverse genome-wide coverage in a high-throughput array format. Designed for inclusive GWAS and imputation studies, PangenomePro helps researchers generate more representative genetic insights across global populations.

The Axiom PangenomePro Plus Array delivers ethnically diverse, genome-wide coverage in a high-throughput 96-array plate format, designed to support whole-genome imputation across global populations and help researchers build more representative genomic datasets. It can serve as a core research tool for human genomics workflows, including GWAS, population health initiatives, polygenic risk scores, pharmacogenomics, and drug discovery.

Features of the Axiom PangenomePro Plus Array include:

  • Marker selection emphasizes broad imputation power across European, African, admixed American, East Asian, and South Asian populations Low input, plug-and-play analysis.
  • Whole-genome analysis from as little as 100 ng genomic DNA with support for target SNP identification, CNV analysis, HLA and blood typing, and 'plug and play' data analysis workflows.
  • GWAS imputation module (~850,000 markers) across ancestral populations
  • Evidence-based clinical/research markers (includes references to public resources such as ACMG 73, ClinVar, NHGRI-GWAS catalog, etc)
  • Pharmacogenomics coverage aligned to guideline and knowledgebase citations (e.g., CPIC, PharmGKB, PharmaADME)
  • Targeted amplification access for challenging/high-homology regions
  • CNV analysis for fixed regions and de novo discovery across the genome

Plus assay advantage

The 'Plus' workflow unlocks pharmacogenomics content and markers for haplotypes in key genes (e.g., CYP2D6) and addresses limitations in other hybridization-based approaches via gene-specific amplification.

Pharmacogenomics Knowledge Base (PharmGKB™)

  • >2,000 markers in Very Important Pharmacogenes as identified by PharmGKB™
  • >300 markers associated with PharmGKB™ level 1A-2B annotations
  • >550 reportable alleles mentioned in Clinical Pharmacogenetics Implementation Consortium (CPIC™) guidelines
  • Targeted amplification to access content in the following genes: CYP1A2, CYP2D6, CYP2B6, CYP2A6, CYP2C19, and those in in highly homologous regions such as CYP2C9, GSTM1, SULT1A1
  • Disease-related variants (Alzheimer's, cancer, cardiovascular disease, cardiometabolic disease, diabetes, neurological disorders)
  • Blood phenotyping variants for blood typing of common and rare blood groups, bleeding disorders, and blood conditions such as sickle cell anemia
  • Human leukocyte antigen (HLA) type of 11 major MHC (major histocompatibility complex) Class I and Class II loci
  • SARS-CoV-2 immune-related markers (human leukocyte antigens, killer immunoglobulin-like receptors, autoimmune markers, and inflammatory markers)
  • Functional variants (loss of function variants, expression quantitative trait loci, nonsynonymous variants, lung phenotypes)
  • Ancestry, wellness, and trait-related variants

Pharmacogenomics Translation Reports

The Pharmacogenomics Translation Reports include:

  • >110 genes
  • >75 genes reporting haplotypes
  • >1000 reportable haplotypes
  • >30 genes with phenotype predictions (e.g., intermediate metabolizer, etc)
TRUSTED_SUSTAINABILITY

Specifications

Type Human Microarray Genotyping
Array Genotyping, DNA, Copy Number
Number of Arrays 1 array
Format 96-array Plate
Species Human
No. of Samples 1x96F
Quantity 96 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.